Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04143

Supelco

Diethylene glycol dimethyl ether

analytical standard

Synonym(s):

diEGdiME, 2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736101
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.6 (vs air)

vapor pressure

3 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

370 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

17.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.408 (lit.)
n20/D 1.408

bp

162 °C (lit.)

mp

−64 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol dimethyl ether is a combustible liquid, which can be used in the synthesis of films, chemically similar to polyethylene glycol on low-density polyethylene and Si(100) surfaces via plasma polymerization and deposition.

Application

Diethylene glycol dimethyl ether may be used as an analytical reference standard for the determination of the analyte in water as well as fruit and vegetable samples by various chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

recommended

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Classifying eluents in reversed-phase thin-layer chromatography by spectral mapping
Cserhati T and Bordas Barna
Journal of Chromatography A, 286(5), 131-137 (1984)
Synthesis of polyethylene glycol-like films from capacitively coupled plasma of diethylene glycol dimethyl ether monomer
Cheng.Q and Komvopoulos.K
The Journal of Physical Chemistry, 113, 213-219 (2008)
Foden.RC and Weddell.LJ
Hazardous Materials: Emergency Action Data (1991)
Evaluation of analyte protectants to improve gas chromatographic analysis of pesticides.
Anastassiades M, et al.
Journal of Chromatography A, 1015(1-2), 163-184 (2003)
Simultaneous determination of six hydrophilic ethers at trace levels using coconut charcoal adsorbent and gas chromatography/mass spectrometry.
Stepien DK and Puttmann W.
Analytical and Bioanalytical Chemistry, 405(5), 1743-1751 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service