Skip to Content
MilliporeSigma

1.07004

Oxford Agar (Base)

GranuCult® prime, according to FDA (BAM), ISO 11290, granular, for Listeria spp., pkg of 500 g

Synonym(s):

Listeria selective agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
GranuCult® prime
Agency:
ISO 11290
Ph:
6.9-7.1 (35 °C, 58.5 g/L in H2O, after autoclaving)
Form:
granular
Suitability:
Listeria spp. (spp.)
Application(s):
food and beverages
microbiology
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 11290

Quality Segment

reg. compliance

according to FDA (BAM)

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

6.9-7.1 (35 °C, 58.5 g/L in H2O, after autoclaving)

solubility

58.5 g/L

bulk density

810 kg/m3

application(s)

food and beverages
microbiology

compatibility

for use with Oxford Listeria Selective Supplement (1.07006)

storage temp.

15-25°C

suitability

Listeria spp. (spp.)

General description

GranuCult® Oxford Agar (Base) formulation is based on Columbia Agar with the addition of lithium chloride, acriflavin, colistin sulfate, cefotetan, cycloheximide, and fosfomycin. These ingredients suppress the growth of the common bacteria (e.g., Gram-negative bacteria and a greater part of Gram-positive bacteria). Lithium chloride is one of the ingredients of Oxford Agar base, while the other substances are derived from the Oxford Listeria Selective Supplement (1.07006). Listeria monocytogenes hydrolyses esculin to esculetin and forms a black complex with iron (III) ions, thereby producing brown-green colored colonies with a black halo.

Application

GranuCult® Oxford Agar (Base) is a selective agar for the isolation and detection of Listeria monocytogenes in milk and milk products and other food products.

Analysis Note

Base tested with Oxford-Listeria-selektiv-supplement art. no. 1.07006.
Appearance (clearness): clear
Appearance (color): blueish-brown to greenish-brown
pH-value (25 °C): 6.8 - 7.2
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Listeria monocytogenes ATCC 35152 (WDCM 00109)):
Inoculum on reference medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)):
Inoculum on reference medium (Listeria innocua ATCC 33090 (WDCM 00017)):
Colony count (Listeria monocytogenes ATCC 35152 (WDCM 00109)):
Colony count (Listeria monocytogenes ATCC 13932 (WDCM 00021)):
Colony count (Listeria innocua ATCC 33090 (WDCM 00017)):
Recovery on test medium (Listeria monocytogenes ATCC 35152 (WDCM 00109)): ≥ 50
Recovery on test medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)): ≥ 50
Recovery on test medium (Listeria innocua ATCC 33090 (WDCM 00017)): ≥ 50
Growth Escherichia coliATCC 25922 (WDCM 00013): total inhibition
Growth(Escherichia coli ATCC 8739 (WDCM 00012)): total inhibition
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): partial inhibition
Growth (Candida albicans ATCC 10231 (WDCM 00054)): partial inhibition
Black zone (Listeria monocytogenes ATCC 35152 (WDCM 00109)): black zone
Black zone (Listeria monocytogenes ATCC 13932 (WDCM 00021)): black zone
Black zone (Listeria innocua ATCC 33090 (WDCM 00017)): black zone
Incubation: up to 48 hours at 35 ± 1 °C, aerobic
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library