Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04100

Sigma-Aldrich

Ethylhydrazine oxalate

≥96.0% (T)

Synonym(s):

Ethylhydrazine ethanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8N2 · C2H2O4
CAS Number:
Molecular Weight:
150.13
Beilstein:
3697941
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (T)

impurities

~2% hydrazine

mp

170-173 °C (dec.)

SMILES string

CCNN.OC(=O)C(O)=O

InChI

1S/C2H8N2.C2H2O4/c1-2-4-3;3-1(4)2(5)6/h4H,2-3H2,1H3;(H,3,4)(H,5,6)

InChI key

DUMHBFMURBWDPC-UHFFFAOYSA-N

Application

Ethylhydrazine oxalate can be used to synthesize:
  • 1-ethyl-1H-indazoles
  • 3-substituted 1-ethyl-6-methylpyrimido[4,5-c]pyridazine-5,7(1H,6H)-dione derivatives
  • 4-(2-alkyl-5-methyl-2H-pyrazol-3-yl)-piperidines

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parallel Synthesis of Novel 3-Substituted 1-Ethyl-6-methylpyrimido [4, 5-c] pyridazine-5, 7 (1 H, 6 H)-dione Analogs.
Chen Y, et al.
Synthetic Communications, 40(6), 821-832 (2010)
Regioselective synthesis of 4-(2-alkyl-5-methyl-2H-pyrazol-3-yl)-piperidines.
Dirat O, et al.
Tetrahedron Letters, 47(11), 1729-1731 (2016)
Giorgio Ramadori et al.
Cell reports, 30(11), 3851-3863 (2020-03-19)
Cancer therapy is limited, in part, by lack of specificity. Thus, identifying molecules that are selectively expressed by, and relevant for, cancer cells is of paramount medical importance. Here, we show that peptidyl-prolyl-cis-trans-isomerase (PPIase) FK506-binding protein 10 (FKBP10)-positive cells are
A method for the regioselective synthesis of 1-alkyl-1H-indazoles.
Liu HJ, et al.
Tetrahedron, 69(19), 3907-3912 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service