Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

B22852

Sigma-Aldrich

Benzylhydrazine dihydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHNH2 · 2HCl
CAS Number:
Molecular Weight:
195.09
Beilstein:
3688990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

143-145 °C (dec.) (lit.)

SMILES string

Cl.Cl.NNCc1ccccc1

InChI

1S/C7H10N2.2ClH/c8-9-6-7-4-2-1-3-5-7;;/h1-5,9H,6,8H2;2*1H

InChI key

MSJHOJKVMMEMNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D J Merkler et al.
Archives of biochemistry and biophysics, 317(1), 93-102 (1995-02-20)
Peptidylglycine alpha-amidating enzyme catalyzes the two-step conversion of C-terminal glycine-extended peptides to C-terminal alpha-amidated peptides and glyoxylate in a reaction that requires O2, ascorbate and 2 mol of copper per mole of enzyme [Kulathila et al. (1994) Arch. Biochem. Biophys.
A Bellelli et al.
European journal of biochemistry, 267(11), 3264-3269 (2000-05-29)
The presteady-state and steady-state kinetics of bovine serum amine oxidase (BSAO) were analyzed by stopped-flow transient spectroscopy. A simplified model of the catalytic cycle was found to describe the experimental data and the rate constants of the individual steps were
L Morpurgo et al.
The Biochemical journal, 260(1), 19-25 (1989-05-15)
Bovine serum amine oxidase is inhibited by benzylhydrazine (BHy), but recovers full activity after a few hours incubation [Hucko-Haas & Reed (1970) Biochem. Biophys. Res. Commun. 38, 396-400]. The first phase of the process, requiring about 15 min, was found
Han-Zhong Zhang et al.
Bioorganic & medicinal chemistry, 12(13), 3649-3655 (2004-06-10)
A series of indole-2-carboxylic acid benzylidene-hydrazides has been identified as a new class of potent apoptosis inducers through a novel cell-based caspase HTS assay. The screening hit, 5-chloro-3-methyl-indole-2-carboxylic acid (4-nitrobenzylidene)-hydrazide (3a), was found to arrest T47D cells in G(2)/M and
F N Bolkenius et al.
Biochemical pharmacology, 53(11), 1695-1702 (1997-06-01)
Peptidylglycine alpha-hydroxylating monooxygenase (PHM; EC 1.14.17.3) catalyses the rate-limiting step in the post-translational activation of substance P, among other neuropeptides, from its glycine-extended precursor. Comparative kinetic studies were performed, using trans-styrylacetic acid or trans-styrylthioacetic acid as known mechanism-based inhibitors, of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service