Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901215

Sigma-Aldrich

Ephos

≥95%

Synonym(s):

Dicyclohexyl(3-isopropoxy-2′,4′,6′-triisopropyl-[1,1′-biphenyl]-2-yl)phosphane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H55OP
CAS Number:
Molecular Weight:
534.80
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

161-162 °C

functional group

phosphine

SMILES string

P(C4CCCCC4)(C3CCCCC3)c1c(cccc1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC(C)C

InChI

1S/C36H55OP/c1-24(2)28-22-32(25(3)4)35(33(23-28)26(5)6)31-20-15-21-34(37-27(7)8)36(31)38(29-16-11-9-12-17-29)30-18-13-10-14-19-30/h15,20-27,29-30H,9-14,16-19H2,1-8H3

InChI key

FDAKNRLXSJBEFL-UHFFFAOYSA-N

Application

Buchwald ligand for Pd-catalyzed C–N cross-coupling between primary amines and aryl
halides to form 2-arylaminooxazoles. Catalyst system was reported with NaOPh (CDS001581).
Ephos can be used as a ligand in the palladium-catalyzed synthesis of 4-arylaminothiazoles and 2-arylaminooxazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic Insight Leads to a Ligand That Facilitates the Pd?Catalyzed Formation of 2?(Hetero) Arylaminooxazoles and 4?(Hetero) Arylaminothiazoles.
Olsen E P, et al.
Angewandte Chemie (International Edition in English) (2017)

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service