All Photos(1)
About This Item
Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Recommended Products
grade
SAJ special grade
form
liquid
reaction suitability
reagent type: oxidant
availability
available only in Japan
concentration
0.01 M
1/20 N
SMILES string
[K][Mn](=O)(=O)(=O)=O
InChI
1S/K.Mn.4O/q+1;;;;;-1
InChI key
VZJVWSHVAAUDKD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jenny Li-Ying Huang et al.
Cancer research, 75(16), 3384-3397 (2015-06-27)
GNAQ and GNA11 are heterotrimeric G protein alpha subunits, which are mutated in a mutually exclusive pattern in most cases of uveal melanoma, one of the most aggressive cancers. Here we introduce the first transgenic mouse model of uveal melanoma
Zhanchao Meng et al.
Nature communications, 6, 6096-6096 (2015-02-05)
Indolosesquiterpenoids are a growing class of natural products that exhibit a wide range of biological activities. Here, we report the total syntheses of xiamycin A and oridamycins A and B, indolosesquiterpenoids isolated from Streptomyces. Two parallel strategies were exploited to
Ming Yang et al.
Nature communications, 6, 6445-6445 (2015-03-12)
Clostrubin is a potent antibiotic against methicillin- and vancomycin-resistant bacteria that was isolated from a strictly anaerobic bacterium Clostridium beijerinckii in 2014. This polyphenol possesses a fully substituted arene moiety on its pentacyclic scaffold, which poses a considerable challenge for
Darryl B Jones et al.
Water research, 46(17), 5491-5498 (2012-08-15)
In this study, the impacts of three preoxidation strategies [i.e., using potassium permanganate (KMnO(4)), chlorine dioxide (ClO(2)), or hydrogen peroxide (H(2)O(2))] before preformed monochloramine (NH(2)Cl) addition on the formation and speciation of iodinated trihalomethanes (I-THMs) were evaluated at the Br(-)/I(-)
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service