Skip to Content
MilliporeSigma

Skip To

1.09074

Sulfuric acid solution

c(H₂SO₄) = 0.05 mol/l (0.1 N), reag. USP, ready-to-use volumetric solution for titration, Titripur®

Synonym(s):

Sulfuric acid solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
NACRES:
NA.21
UNSPSC Code:
12352301
MDL number:
Beilstein/REAXYS Number:
2037554
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sulfuric acid, c(H2SO4) = 0.05 mol/l (0.1 N), Titripur®, reag. USP

Quality Segment

agency

reag. USP, suitable for SM 2310, suitable for SM 2320

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

packaging

Titripac® of

concentration

0.05 M

technique(s)

titration: suitable

pH

1.3 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Application


  • Cellulose Nanocrystals and Lignin Nanoparticles Extraction from Lemna minor L.: Acid Hydrolysis of Bleached and Ionic Liquid-Treated Biomass.: This research explores the extraction of cellulose nanocrystals and lignin nanoparticles using sulfuric acid hydrolysis, highlighting its significance in developing sustainable biomaterials (Puglia et al., 2024).

  • Production and Characterization of Nanocellulose from Maguey (Agave cantala) Fiber.: The study utilizes sulfuric acid for the production and characterization of nanocellulose from Maguey fiber, underscoring its potential in advancing biocomposites (Sumarago et al., 2024).

  • Stability and Structure of Potentially Atmospherically Relevant Glycine Ammonium Bisulfate Clusters.: This paper investigates the stability and structure of glycine ammonium bisulfate clusters, providing insights into atmospheric chemistry and the role of sulfuric acid in aerosol formation (Hariharan et al., 2024).

  • Nanostructured Niobium and Titanium Carbonitrides as Electrocatalyst Supports.: Examines the use of sulfuric acid in synthesizing nanostructured niobium and titanium carbonitrides, essential for improving electrocatalyst supports in energy applications (McLeod et al., 2024).

  • Synthesis and characterization of hydroxyapatite nanoparticles from calcium hydroxide fouled with gases evolved from smokestack of glass industry.: This research focuses on the synthesis of hydroxyapatite nanoparticles using sulfuric acid, showcasing its utility in recycling industrial waste (Abdelmoaty & Mousa, 2024).

Features and Benefits

Digital Titration

ChemiSphere™ App for Digital Data Transfer to OMNIS Titration Instruments from Metrohm

Discover the ChemiSphere™ App for secure, rapid data transfer to OMNIS titrators, enhancing efficiency in the titration workflow with Titripur® and Aquastar® reagents. ChemiSphere App for Digital Reagent Data Transfer



SmartChemicals for Digital Data Transfer to Mettler Toledo Titration Instruments

Explore digital data transfer with our SmartChemicals for Mettler Toledo titrators—fast, easy, and safe, accelerating the titration process with Titripur®, Aquastar® reagents, and Certipur® and Aquastar® standards. Supelco® SmartChemicals for Titration
Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Titripac® packaging - all SKU-pack size numbers ending "4000, 4003, 9010, 9013" - more information on www.sigmaaldrich.com/Titripac

Analysis Note

Form liquid
Amount-of-substance concentration 0.04975 - 0.05025 mol/L
Measurement uncertainty ± 0.00015 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 723 Tris(hydroxymethyl)aminomethane) by means of volumetric standard Tris(hydroxymethyl)aminomethane (article number 1.02408), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Other Notes

Replaces: SX1243G-6; SX1243G-3

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
ChemiSphere is a trademark of Merch KGaA, Darmstadt, Germany
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Sulfuric acid


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters