Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.60313

Supelco

Sulfuric acid solution

c(H₂SO₄) = 1 mol/l (2 N), ready-to-use volumetric solution for titration, Titriplex®

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352322
NACRES:
NA.21

product name

Sulfuric acid, c(H2SO4) = 1 mol/l (2 N), Titripur®

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

concentration

1 M

technique(s)

titration: suitable

pH

1 (20 °C in H2O)

density

1.1 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.995 - 1.005 mol/L
Measurement uncertainty ± 0.003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 723 Tris(hydroxymethyl)aminomethane) by means of volumetric standard Tris(hydroxymethyl)aminomethane (article number 1.02408), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service