Skip to Content
MilliporeSigma
All Photos(1)

Documents

23-0590

Sigma-Aldrich

Oleic acid

SAJ first grade, ≥70.0%

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 176 °C)

Assay

≥70.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

oleic acid

storage temp.

room temp

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joyce Wang et al.
BMC genomics, 15, 415-415 (2014-06-03)
Mycobacterium avium subsp. paratuberculosis (MAP) is an obligate intracellular pathogen that infects many ruminant species. The acquisition of foreign genes via horizontal gene transfer has been postulated to contribute to its pathogenesis, as these genetic elements are absent from its
Marta Estrader et al.
Nanoscale, 7(7), 3002-3015 (2015-01-21)
The intimate relationship between stoichiometry and physicochemical properties in transition-metal oxides makes them appealing as tunable materials. These features become exacerbated when dealing with nanostructures. However, due to the complexity of nanoscale materials, establishing a distinct relationship between structure-morphology and
Stefan Freigang et al.
Nature immunology, 14(10), 1045-1053 (2013-09-03)
Chronic inflammation is a fundamental aspect of metabolic disorders such as obesity, diabetes and cardiovascular disease. Cholesterol crystals are metabolic signals that trigger sterile inflammation in atherosclerosis, presumably by activating inflammasomes for IL-1β production. We found here that atherogenesis was
Ying Liu et al.
American journal of physiology. Renal physiology, 305(11), F1555-F1562 (2013-08-16)
Diabetic nephropathy (DN) is a leading cause of end-stage renal disease (ESRD). The inhibitors of renin-angiotensin-aldosterone system (RAAS) can alleviate some of the symptoms of DN but fail to stop the progression to ESRD. Our previous studies demonstrate renoprotective action
Wim Reybroeck et al.
PloS one, 9(5), e98266-e98266 (2014-05-24)
Two Pseudomonas strains, identified as closely related to Pseudomonas tolaasii, were isolated from milk of a farm with frequent false-positive Delvotest results for screening putative antibiotic residues in raw milk executed as part of the regulatory quality programme. Growth at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service