Skip to Content
MilliporeSigma
All Photos(1)

Documents

N4876

Sigma-Aldrich

Ninhydrin

suitable for amino acid detection

Synonym(s):

1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein:
1910963
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

form

powder

Quality Level

mp

250 °C (dec.) (lit.)

solubility

ethanol: 100 mg/mL
H2O: 50 mg/mL (with sonication)

suitability

suitable for amino acid detection

SMILES string

OC1(O)C(=O)c2ccccc2C1=O

InChI

1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H

InChI key

FEMOMIGRRWSMCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ninhydrin is an important building block in organic synthesis and has been employed to prepare a variety of chemicals for complexing and/or sensing metal ions. It is an amino acid-sensitive reagent used for the preparation of Ruhemann′s purple.

Application

Ninhydrin can be used:
  • For the detection of free amino groups in amino acids, peptides, and proteins.
  • As a ligand for the Cu-catalyzed N-arylation of nitrogen-containing heterocycles with aryl and heteroaryl halides to synthesize N-arylheterocycles.
  • As a reactant to synthesize aldols by reacting with active methylene compounds.
Used for the detection of free amino groups in amino acids, peptides and proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reaction of diaminofurazane with ninhydrin.
Pirogov SV, et al.
Chemistry of Heterocyclic Compounds, 33(12), 1473-1474 (1997)
Vibrational spectroscopy and density functional theory study of ninhydrin.
Li R, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 136, 1642-1648 (2015)
Quantitative determination of chitosans by ninhydrin.
Prochazkova S, et al.
Carbohydrate Polymers, 38(2), 115-122 (1999)
Substituted naphthoquinones as novel amino acid sensitive reagents for the detection of latent fingermarks on paper surfaces
Jelly R, et al.
Talanta, 82, 1717-1724 (2010)
Isolation and activation of collagenase from fish processing waste.
Daboor SM, et al.
Advances in Bioscience and Biotechnology, 3(3) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service