Skip to Content
MilliporeSigma
All Photos(1)

Documents

15-2320

Sigma-Aldrich

2-Propanol

≥99.5%, JIS special grade

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

product name

2-Propanol, JIS special grade, ≥99.5%

grade

JIS special grade

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

availability

available only in Japan

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.1 °F

Flash Point(C)

11.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter C Grayson et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(7), 1922-1932 (2015-04-22)
To discover biomarkers involved in the pathophysiology of antineutrophil cytoplasmic antibody-associated vasculitis (AAV) and to determine whether low-density granulocytes (LDGs) contribute to gene expression signatures in AAV. The source of clinical data and linked biologic specimens was a randomized controlled
Jan Rybniker et al.
Nature communications, 6, 7659-7659 (2015-07-15)
Better antibiotics capable of killing multi-drug-resistant Mycobacterium tuberculosis are urgently needed. Despite extensive drug discovery efforts, only a few promising candidates are on the horizon and alternative screening protocols are required. Here, by testing a panel of FDA-approved drugs in
H Dolatshad et al.
Leukemia, 29(5), 1092-1103 (2014-11-28)
The splicing factor SF3B1 is the most commonly mutated gene in the myelodysplastic syndrome (MDS), particularly in patients with refractory anemia with ring sideroblasts (RARS). We investigated the functional effects of SF3B1 disruption in myeloid cell lines: SF3B1 knockdown resulted
Wei-Yu Lin et al.
Human molecular genetics, 24(1), 285-298 (2014-08-30)
Previous studies have suggested that polymorphisms in CASP8 on chromosome 2 are associated with breast cancer risk. To clarify the role of CASP8 in breast cancer susceptibility, we carried out dense genotyping of this region in the Breast Cancer Association
Emilie Montastier et al.
PLoS computational biology, 11(1), e1004047-e1004047 (2015-01-16)
Nutrigenomics investigates relationships between nutrients and all genome-encoded molecular entities. This holistic approach requires systems biology to scrutinize the effects of diet on tissue biology. To decipher the adipose tissue (AT) response to diet induced weight changes we focused on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service