Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1457833

USP

Nebivolol hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Narbivolol, Nebilet, R-65824, rel-(aR,a′R,2R,2′S)-a,a′-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol] hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25F2NO4 · HCl
CAS Number:
Molecular Weight:
441.90
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nebivolol

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Cl.O[C@@H](CNC[C@H](O)[C@@H]1CCc2cc(F)ccc2O1)[C@H]3CCc4cc(F)ccc4O3

InChI

1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m0./s1

InChI key

JWEXHQAEWHKGCW-BIISKSHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nebivolol hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Nebivolol is a competitive, highly selective b1-receptor antagonist with mild vasodialating properties, possibly due to an interaction with the L-arginine/ NO pathway, and is used for treatment of essential hypertension.
Nebivolol is a competitive, highly selective b1-receptor antagonist with mild vasodialating properties, possibly due to an interaction with the L-arginine/ NO pathway, and is used for treatment of essential hypertension. It is 40-fold selective for β1 vs. β2, and lacks ISA (intrinsic sympathomimetic activity).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service