Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-2360

Sigma-Aldrich

2-Propanol

≥99.7% (GC), suitable for HPLC

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
51471901
PubChem Substance ID:
Assay:
≥99.7% (GC)
technique(s):
HPLC: suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, suitable for HPLC

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.1 °F

Flash Point(C)

11.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter C Grayson et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(7), 1922-1932 (2015-04-22)
To discover biomarkers involved in the pathophysiology of antineutrophil cytoplasmic antibody-associated vasculitis (AAV) and to determine whether low-density granulocytes (LDGs) contribute to gene expression signatures in AAV. The source of clinical data and linked biologic specimens was a randomized controlled
Jan Rybniker et al.
Nature communications, 6, 7659-7659 (2015-07-15)
Better antibiotics capable of killing multi-drug-resistant Mycobacterium tuberculosis are urgently needed. Despite extensive drug discovery efforts, only a few promising candidates are on the horizon and alternative screening protocols are required. Here, by testing a panel of FDA-approved drugs in
H Dolatshad et al.
Leukemia, 29(5), 1092-1103 (2014-11-28)
The splicing factor SF3B1 is the most commonly mutated gene in the myelodysplastic syndrome (MDS), particularly in patients with refractory anemia with ring sideroblasts (RARS). We investigated the functional effects of SF3B1 disruption in myeloid cell lines: SF3B1 knockdown resulted
Thomas W Winkler et al.
PLoS genetics, 11(10), e1005378-e1005378 (2015-10-02)
Genome-wide association studies (GWAS) have identified more than 100 genetic variants contributing to BMI, a measure of body size, or waist-to-hip ratio (adjusted for BMI, WHRadjBMI), a measure of body shape. Body size and shape change as people grow older
Swapna Ganapathy et al.
Journal of the American Chemical Society, 136(46), 16335-16344 (2014-10-24)
Fundamental research into the Li-O2 battery system has gone into high gear, gaining momentum because of its very high theoretical specific energy. Much progress has been made toward understanding the discharge mechanism, but the mechanism of the oxygen evolution reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service