Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-0620

Sigma-Aldrich

Iodine solution

0.05 M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:

availability

available only in Japan

concentration

0.05 M
1/10 N

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

O Barbier et al.
The Knee, 22(5), 380-383 (2015-06-07)
Inadvertent contamination of the autograft can occur during anterior cruciate ligament (ACL) reconstruction if the autograft is dropped on the floor during surgery. A study was undertaken to determine the incidence of contamination when a graft is dropped on the
Hyo-Jung Lee et al.
Journal of periodontal & implant science, 45(3), 94-100 (2015-07-02)
The aim of this study was to investigate the combined effects of physical and chemical surface factors on in vivo bone responses by comparing chemically modified hydrophilic sandblasted, large-grit, acid-etched (modSLA) and anodically oxidized hydrophobic implant surfaces. Five modSLA implants
Yanfeng Zhang et al.
Experimental and therapeutic medicine, 9(4), 1351-1356 (2015-03-18)
Oxytocin (OT) neurons localized in the paraventricular nucleus (PVN) and supraoptic nucleus (SON) send fibers to the brain and spinal cord. While most previous studies have looked at the role of OT in chronic pain, few have investigated the role
Ü Yolcu et al.
International journal of oral and maxillofacial surgery, 44(11), 1390-1397 (2015-08-10)
The aim of this study is to introduce a new flap design in the surgical removal of impacted mandibular third molars - a lingually based triangular flap - and to compare this flap design with the routinely used triangular flap.
Hélène Guet-Revillet et al.
Emerging infectious diseases, 20(12), 1990-1998 (2014-11-25)
Hidradenitis suppurativa (HS) is a skin disease characterized by recurrent nodules or abscesses and chronic suppurating lesions. In the absence of clear pathophysiology, HS is considered to be an inflammatory disease and has no satisfactory medical treatment. Recently, prolonged antimicrobial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service