Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09910

Supelco

Iodine solution

c(I₂) = 0.005 mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141916
NACRES:
NA.21

product name

Iodine solution,

pH

6.7 (20 °C in H2O)

Quality Level

density

1.38 g/cm3 at 20 °C

storage temp.

20-25°C

General description

for 1000 ml, c(I₂) = 0.05 mol/l (0.1 N) Titrisol®

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service