Skip to Content
MilliporeSigma

IX0130A

Iodine solution

c(I₂) = 0.5 mol/l (1 N)

Synonym(s):

normality

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12141916
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Iodine solution (0.1N),

agency

traceable to NIST

Quality Segment

form

liquid

shipped in

ambient

storage temp.

room temp



Still not finding the right product?

Explore all of our products under Iodine solution (0.1N)


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2 Oral

target_organs

Thyroid

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library