Skip to Content
MilliporeSigma
All Photos(1)

Documents

12-1541

Sigma-Aldrich

Gram-Hucker′s stain solution (III)

Synonym(s):

Safranin T solution, Gram’s safranin solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Beilstein:
3924099
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

availability

available only in Japan

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Náglio Kalil Dias et al.
Connective tissue research, 53(1), 39-47 (2011-09-22)
The aim of study was to evaluate the progression of the ankle articular cartilage alterations after a post-immobilization muscle stretching. Twenty-nine Wistar rats were separated into five groups: C--control, S--stretched, SR--stretch recovery, IS--immobilized and stretched, and ISR--immobilized stretched recovery. The
Xinlin Yang et al.
Tissue engineering. Part A, 17(11-12), 1595-1603 (2011-02-10)
Tissue engineering is a promising approach for treatment of disc degeneration. Herein, we evaluated effects of rotating bioreactor culture on the extracellular matrix production and proliferation of human annulus fibrosus (AF) cells. AF cells were embedded into alginate beads, and
Hatice T Sanal et al.
Journal of orofacial pain, 25(4), 345-353 (2012-01-17)
To use the ultrashort time-to-echo magnetic resonance imaging (UTE MRI) technique to quantify short T2* properties (obtained through gradient echo) of a disc from the human temporomandibular joint (TMJ) and to corroborate regional T2* values with biomechanical properties and histologic
J E Gil et al.
International journal for numerical methods in biomedical engineering, 28(12), 1184-1197 (2012-12-06)
This work describes an expert system aimed to an accurate classification of cell tissue on microscopic images coming from studies of bone tissue regeneration from stem cells. We analyze a wide number of phenotype and color issues to build effective
Manos Stefanakis et al.
Spine, 37(22), 1883-1891 (2012-06-19)
Mechanical and biochemical analyses of cadaveric and surgically removed discs. To test the hypothesis that fissures in the annulus of degenerated human discs are mechanically and chemically conducive to the ingrowth of nerves and blood vessels. Discogenic back pain is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service