MilliporeSigma
  • Enhancement of matrix production and cell proliferation in human annulus cells under bioreactor culture.
Enhancement of matrix production and cell proliferation in human annulus cells under bioreactor culture.
Tissue engineering. Part A (2011-02-10)
Xinlin Yang, Daidong Wang, Jianrong Hao, Meiqing Gong, Vincent Arlet, Gary Balian, Francis H Shen, Xudong Joshua Li
ABSTRACT
Tissue engineering is a promising approach for treatment of disc degeneration. Herein, we evaluated effects of rotating bioreactor culture on the extracellular matrix production and proliferation of human annulus fibrosus (AF) cells. AF cells were embedded into alginate beads, and then cultured up to 3 weeks in a rotating wall vessel bioreactor or a static vessel. By real-time reverse transcription-polymerase chain reaction, expression of aggrecan, collagen type I and type II, and collagen prolyl 4-hydroxylase II was remarkably elevated, whereas expression of matrix metalloproteinase 3 and a disintegrin and metalloproteinase with thrombospondin motifs 5 was significantly decreased under bioreactor. Biochemical analysis revealed that the levels of the whole cell-associated proteoglycan and collagen were approximately five- and twofolds in rotating bioreactor, respectively, compared to those in static culture. Moreover, AF cell proliferation was augmented in rotating bioreactor. DNA contents were threefolds higher in rotating bioreactor than that in static culture. Expression of the proliferating cell nuclear antigen was robustly enhanced in rotating bioreactor as early as 1 week. Our findings suggested that rotating bioreactor culture would be an effective technique for expansion of human annulus cells for tissue engineering driven treatment of disc degeneration.
Sigma-Aldrich
Gram′s safranin solution, for microscopy
Sigma-Aldrich
Gram-Hucker′s stain solution (III)
Sigma-Aldrich
Safranin O, for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0)
Sigma-Aldrich
Safranin O, certified by the Biological Stain Commission
Sigma-Aldrich
Safranin O, Dye content ≥85 %