Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-4110

Sigma-Aldrich

Dimethylamine solution

CP, 50% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

CP

form

liquid

availability

available only in Japan

concentration

50% in H2O

pH

14

density

0.824 g/cm3

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Mulder et al.
Journal of neural transmission (Vienna, Austria : 1996), 109(9), 1203-1208 (2002-08-31)
Nitric oxide (NO) may play a role in the pathophysiology of Alzheimer's disease (AD). Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of NO synthase, is involved in regulation of NO production. Recently it has been reported that dimethylarginine dimethylaminohydrolase, an enzyme
Gerhild Zauner et al.
Biochimica et biophysica acta, 1820(9), 1420-1428 (2011-08-02)
Analysis of protein glycosylation is an important first step towards establishing the functions of glycans in health and disease. In contrast to N-glycans which are generally enzymatically released for analysis, there is no corresponding enzyme for O-glycan liberation. Therefore, O-glycans
L Lee et al.
Cancer research, 41(10), 3992-3994 (1981-10-01)
Using a method for nitrosamine analysis that gives high recovery values and that is free from artifactual synthesis of nitrosamines, we have shown that human feces do not contain volatile nitrosamines (detection limit, 0.1 to 0.5 microgram/kg). We also showed
Meric Selbes et al.
Water research, 47(2), 945-953 (2012-12-15)
N-nitrosodimethylamine (NDMA), a probable human carcinogen, is a disinfection by-product that has been detected in chloraminated and chlorinated drinking waters and wastewaters. Formation mechanisms and precursors of NDMA are still not well understood. The main objectives of this study were
George Nicholson et al.
PLoS genetics, 7(9), e1002270-e1002270 (2011-09-21)
We have performed a metabolite quantitative trait locus (mQTL) study of the (1)H nuclear magnetic resonance spectroscopy ((1)H NMR) metabolome in humans, building on recent targeted knowledge of genetic drivers of metabolic regulation. Urine and plasma samples were collected from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service