Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.22033

Sigma-Aldrich

Dimethylamine

(40% aqueous solution) for synthesis

Synonym(s):

Dimethylamine solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

vapor pressure

263 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

415 °C

potency

1000 mg/kg LD50, oral (Rat)
3900 mg/kg LD50, skin (Rat)

expl. lim.

2.6-12.3 % (v/v)

pH

14 ( in H2O)

mp

-38 °C

transition temp

flash point -18.5 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Interaction with Oxalic Acid: Discusses the atmospheric implications of the interaction between oxalic acid and dimethylamine, highlighting the formation of complexes and proton transfer events (Chen et al., 2017).
  • Hydroaminoalkylation Reactions: Describes the application of dimethylamine as a substrate in catalytic hydroaminoalkylation reactions, providing a pathway for the synthesis of complex organic compounds (Bielefeld and Doye, 2017).

Analysis Note

Assay (acidimetric): 39.0 - 41.0 %
Density (d 20 °C/ 4 °C): 0.890 - 0.900

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-1.3 °F

Flash Point(C)

-18.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service