Skip to Content
MilliporeSigma
All Photos(1)

Documents

L6155

Sigma-Aldrich

Latex beads, carboxylate-modified, streptavidin-labeled, blue

buffered aqueous suspension, 0.25 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

form

buffered aqueous suspension

composition

Solids, 1.0%

mean particle size

0.25 μm

Looking for similar products? Visit Product Comparison Guide

Application

Carboxylate-modified polystyrene latex beads have been used to collect morphofunctional data about the immune systems of Carabus lefebvrei, a helicophagous Italian endemic ground beetle often used as an indicator of the habitat quality of the Apennines mountain forests. Carboxylate-modified polystyrene latex beads have also been used to inform comparative studies on immunity defense mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa and to investigate the phagocytic activity of cultured rat thyroid cells.

Physical form

Suspension in 10 mM PBS, pH 7.1, 2 mM NaN3

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Courtney Voss et al.
JCI insight, 6(13) (2021-06-04)
BACKGROUNDThe role of humoral immunity in COVID-19 is not fully understood, owing, in large part, to the complexity of antibodies produced in response to the SARS-CoV-2 infection. There is a pressing need for serology tests to assess patient-specific antibody response
Circulating hemocytes from larvae and adults of Carabus (Chaetocarabus) lefebvrei Dejean 1826 (Coleoptera, Carabidae): cell types and their role in phagocytosis after in vivo artificial non-self-challenge
Giglio, A., et al.
Micron (Oxford, England : 1993), 39, 52-58 (2008)
P G Giulianini et al.
Tissue & cell, 35(4), 243-251 (2003-08-19)
In the context of comparative studies on immunity defence mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa (Drury, 1770) the ultrastructure of the circulating hemocytes of the third instar larval stage has been investigated by means of light
K Takazawa et al.
Acta endocrinologica, 117(2), 198-204 (1988-02-01)
We investigated the phagocytic activity of cultured rat thyroid cells (FRTL) by using latex beads. FRTL cells ingested many carboxylate-modified polystyrene latex beads (c-latex beads), which had carboxyl residues on the surface, whereas they ingested few polystyrene latex beads (p-latex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service