Skip to Content
MilliporeSigma
All Photos(1)

Documents

16-126

Millipore

Streptavidin, agarose conjugate

Streptavidin, agarose beads suitable for purification of biotinylated macromolecules.

Synonym(s):

Streptavidin agarose beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32160801
NACRES:
NA.56

form

liquid

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

shipped in

wet ice

General description

Streptavidin isolated from Streptomyces avidinii covalently bound to agarose.

Application

Streptavidin, agarose conjugate has been used in:
  • immunoprecipitation
  • coimmunoprecipitation
  • reversible biotinylation assay
It is suitable for the purification of biotinylated macromolecules.

Quality

routinely evaluated by immunoprecipitating tyrosine-phosphorylated proteins from a RIPA lysate prepared from EGF-stimulated A431 cells with a biotinylated monoclonal phosphotyrosine antibody (16-103)

Physical form

phosphate-buffered saline containing 0.02% sodium azide

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service