Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DPNI-RO

Roche

Dpn I

from Diplococcus pneumoniae

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

bacterial (Diplococcus pneumoniae)

Quality Level

form

solution

packaging

pkg of 1,000 U (10742988001 [10 U/μl])
pkg of 200 U (10742970001 [10 U/μl])

manufacturer/tradename

Roche

parameter

37 °C optimum reaction temp.

storage temp.

−20°C

General description

Isoschizomers
The enzyme is an isoschizomer to Bsp143 I, Dpn II, Mbo I, Nde II and Sau3A I.
Methylation sensitivity
The enzyme is only inhibited by the occurrence of 5-methylcytosine at the indicated site (*) if no 6-methyladenine is present.
Typical ligation and recutting assay
Dpn I fragments obtained by complete digestion of 1μg pBR322 DNA are ligated for 16 hours at +4°C with 1U T4 DNA Ligase in 10μl buffer that contains 66mM Tris-HCl, 5mM MgCl2, 5mM Dithiothreitol, 1mM ATP, pH 7.5 (at +20°C). The percentages of product that can be ligated and subsequently recut with Dpn I (yielding the typical pattern of pBR322 × Dpn I fragments) are stated under "Lig" and "Rec" in the certificate of analysis.
Dpn I requires methylation of adenine residues for activity and thus digests only GmATC sequences containing N6-methyladenine. Such methylated sequences are produced by dam methylase. This requirement for methylation distinguishes Dpn I from Mbo I, which is inhibited by dam methylation, and Sau3A I, which is not influenced by dam methylation. Dpn I also cleaves at a different position on the recognition sequence than Mbo I and Sau3A I.

Specificity

Recognition sites: GmAT*C
GmAT*C
Restriction site: GmA↓T*C
GmA↓T*C
Heat inactivation: No inactivation of Dpn I after incubation at 65 °C for 15 minutes.

Application

In PCR-mediated mutagenesis, DpnI nuclease has been used for the digestion of wild type DNA after PCR.

Quality

Absence of nonspecific endonuclease activities
1μg pBR322 DNA is incubated for 16 hours in 50μl SuRE/Cut Buffer A with an excess of Dpn I. The number of enzyme units which do not change the enzyme-specific pattern is stated in the certificate of analysis.
Absence of exonuclease activity
Approximately 5μg [3H] labeled calf thymus DNA are incubated with 3μl Dpn I for 4 hours at +37°C in a total volume of 100μl 50mM Tris-HCl, 10mM MgCl2, 1mM Dithioerythritol, pH approximately 7.5. Under these conditions, no release of radioactivity is detectable, as stated in the certificate of analysis.

Compatibility

Dpn I generates fragments with blunt ends that are compatible with any blunt end.

DNA Profile

Number of cleavage sites on different DNAs
  • λ: 116
  • φX174: 0
  • Ad2: 87
  • M13mp7: 8
  • pBR322: 22
  • pBR328: 27
  • pUC18: 15
  • SV40: 8

Unit Definition

One unit is the enzyme activity that cleaves 1 μg pBR322 DNA in one hour at +37 °C in a total volume of 25 μl (1x) SuRE/Cut Buffer A. Since complete Dpn I digestion of pBR322 DNA needs fully methylated GATC sequences < 5% partial digested bands may be observed during activity determination.

Analysis Note

SuRE/Cut Buffer System
The buffer in bold is recommended for optimal activity
  • A: 100%
  • B: 75-100%
  • H: 75-100%
  • L: 50-75%
  • M: 75-100%

Activity in PCR buffer: 100%

Relative activity in PCR mix (Taq DNA Polymerase buffer) is 100%. The PCR mix contained λDNA, primers, 10 mM Tris-HCl (pH 8.3, 20 °C), 50 mM KCl, 1.5 mM MgCl2, 200 μM dNTPs, 2.5 U Taq DNA polymerase. The mix was subjected to 25 amplification cycles.Activity in reaction buffer of Pwo SuperYield DNA Polymerase PCR Mix is 100%; Pwo SuperYield DNA Polymerase PCR Mix is not available in U.S.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Enzyme Solution

  • SuRE/Cut Buffer A 10x concentrated

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiyuki Adachi et al.
Infection and immunity, 72(7), 4159-4171 (2004-06-24)
Dectin 1 is a mammalian cell surface receptor for (1-->3)-beta-d-glucans. Since (1-->3)-beta-d-glucans are commonly present on fungal cell walls, it has been suggested that dectin 1 is important for recognizing fungal invasion. In this study we tried to deduce the

Articles

The term “Restriction enzyme” originated from the studies of Enterobacteria phage λ (lambda phage) in the laboratories of Werner Arber and Matthew Meselson.

Related Content

Restriction endonucleases in prokaryotes function primarily to protect against foreign genetic material, notably bacteriophage DNA.

Restriction endonucleases in prokaryotes function primarily to protect against foreign genetic material, notably bacteriophage DNA.

Restriction endonucleases in prokaryotes function primarily to protect against foreign genetic material, notably bacteriophage DNA.

Restriction endonucleases in prokaryotes function primarily to protect against foreign genetic material, notably bacteriophage DNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service