Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS4871

Corning® Costar® reagent reservoirs

white polystyrene, capacity 50 mL

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.14

material

U-bottom bottom
white polystyrene

sterility

sterile

packaging

pack of 100 ea (Individually wrapped)

manufacturer/tradename

Corning 4871

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Costar® reagent disposable reservoirs are used to repetitively fill multichannel pipettors. These reserviors are non-pyrogenic and free of DNase/RNase contamination.

Application

Corning® Costar® reagent reservoirs has been used as a container to hold the parasite inoculum (gametocyte culture) before it is dispensed into the 384-well plates.

Features and Benefits

  • Made from high-quality, virgin white polystyrene material.
  • Each reagent reservoir is individually packaged and provided in a sterile condition
  • The reservoirs are disposable

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Delves et al.
Nature protocols, 11(9), 1668-1680 (2016-08-26)
The prevention of parasite transmission from the human host to the mosquito has been recognized as a vital tool for malaria eradication campaigns. However, transmission-blocking antimalarial drug and/or vaccine discovery and development is currently hampered by the expense and difficulty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service