Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

XX1012542

Millipore

Millipore Glass Base and Stopper for Vacuum Filtration

Synonym(s):

Glass Base for Filtration, Millipore Vacuum Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

parylene coating (on stopper)
silicone stopper
stainless steel base
stainless steel screen

technique(s)

vacuum filtration: suitable

diam.

25 mm

filtration area

2.8 cm2

application(s)

filtration

shipped in

ambient

General description

Millipore Glass Vacuum Filtration Accessory Product
Glass Base and Stopper, 25 mm, Stainless Steel Support, Screen provided.

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 25 mm Analytical Stainless Steel Filter Holder Kit.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service