Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

XX1012532

Millipore

Millipore Glass Base and Stopper for Vacuum Filtration

25 mm, For use with stainless steel screen support, Screen provided

Synonym(s):

Glass stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

borosilicate glass
parylene coating (on stopper)
silicone stopper
stainless steel screen

technique(s)

vacuum filtration: suitable

diam.

25 mm

application(s)

filtration

shipped in

ambient

General description

Millipore Glass Vacuum Filtration Accessory Product
Glass Base and Stopper, 25 mm, For use with stainless steel screen support, Screen provided

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service