Skip to Content
MilliporeSigma
All Photos(1)

Documents

WBLUF

Millipore

Immobilon Forte Western HRP substrate, 100 mL

Western blotting, dot/slot blotting

Synonym(s):

HRP substrate, chemiluminescent detection substrate, detection substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

conjugate

peroxidase conjugate

Quality Level

packaging

pkg of 100 mL

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

membrane area

1000 cm2 , membrane coverage

detection method

chemiluminescent

shipped in

ambient

General description

Premixed, ready to use chemiluminescent HRP detection reagent

Application

For use on immunoblots.
Western blotting, dot/slot blotting

Packaging

1 bottle for 1000 cm2

Quality

The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.

Storage and Stability

Stable for 1 year at room temperature from date of shipment. Contains hydrogen peroxide. Use in well ventilated area.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Comparison of elution techniques for small-scale protein purification of FLAG® tag proteins using anti-FLAG® M2 magnetic beads.

A discussion of factors affecting observed vs. calculated molecular weight in gel electrophoresis and western blotting.

Protocols

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service