Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.83
UNSPSC Code:
12352204
Application
One Component Chemiluminescent Membrane / Blotting Peroxidase Substrate is luminol-based and can detect horseradish peroxidase at high sensitivity levels (low picogram to femtogram). It provides superior sensitivity and convenience compared to competitor products. The substrate is supplied as one component. It requires no mixing and is ready to use, thus eliminating mixing errors. CPSOC substrate may be used for immunoblots, western blots, dot blots and any blotting application utilizing horseradish peroxidase (HRP)-conjugates. The substrate can be used with various blocking buffers and on nitrocellulose or PVDF membranes. Such blots will exhibit low backgrounds. Detection and analysis may be done by CCD imaging systems or x-ray film.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.