Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R0278

Sigma-Aldrich

RIPA Buffer

RIPA Buffer

Synonym(s):

Radioimmunoprecipitation assay buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161703
NACRES:
NA.56
Pricing and availability is not currently available.

description

protease free

Quality Level

form

liquid

packaging

pkg of 50 mL
pkg of 500 mL

technique(s)

immunoprecipitation (IP): suitable

color

colorless

pH

7.8-8.2 (25 °C, 1 ×)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RIPA (Radio-Immunoprecipitation Assay) Buffer is supplied as a ready to use solution that requires no preparation. It minimizes non-specific protein-binding interactions to keep background low, while allowing most specific interactions to occur, enabling studies of relevant protein-protein interactions. It is widely used in applications such as immunoprecipitation since most antibodies and protein antigens are not adversely affected by the components of this buffer.
Radioimmunoprecipitation assay buffer (RIPA buffer) is a lysis buffer used for rapid, efficient cell lysis and solubilization of proteins from both adherent and suspension cultured mammalian cells.

Application

RIPA Buffer enables efficient cell lysis and protein solubilization while avoiding protein degradation and interference with the proteins′ immunoreactivity and biological activity. RIPA Buffer also results in low background in immunoprecipitation and molecular pull-down assays. Compatible with EZview Affinity Gels.
RIPA Buffer has been used as a reagent for cell lysis in the following experimental studies:
  • Fibroblast-based deep phenotyping of the peripheral tissue, which facilitates the mechanistic disease stratification in sporadic Parkinson′s disease (sPD).[1]
  • Demonstrating the inhibition of oxidative phosphorylation (OXPHOS), which can activate the mixed lineage kinase domain-like protein (MLKL)-dependent necroptosis in human lung epithelial cells.[2]

Features and Benefits

1 mL of the RIPA Buffer is used to lyse cells from one 100 mm culture dish (0.5 to 5 * 107 cells) of most adherent mammalian cell lines.
Minimizes non-specific protein binding interactions to ensure low background.

Other Notes

Ready-to-use solution containing 150 mM NaCl, 1.0% IGEPAL® CA-630, 0.5% sodium deoxycholate, 0.1% SDS, 50 mM Tris, pH 8.0.

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC
IGEPAL is a registered trademark of Solvay

also commonly purchased with this product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elham Abbasloo et al.
Neuropeptides, 60, 7-12 (2016-08-21)
Pain represents a major contributing factor to the individual's quality of life. Although pain killers as opioids, endogenous or exogenous peptides can decrease pain perception, the chronic use of them leads to antinociceptive tolerance. It has been demonstrated that neuropeptide
Zuzana Keckesova et al.
Nature, 543(7647), 681-686 (2017-03-23)
Post-mitotic, differentiated cells exhibit a variety of characteristics that contrast with those of actively growing neoplastic cells, such as the expression of cell-cycle inhibitors and differentiation factors. We hypothesized that the gene expression profiles of these differentiated cells could reveal
Hartmut Jahns et al.
Nature communications, 6, 6317-6317 (2015-03-07)
An established means of improving the pharmacokinetics properties of oligoribonucleotides (ORNs) is to exchange their phosphodiester linkages for phosphorothioates (PSs). However, this strategy has not been pursued for small interfering RNAs (siRNAs), possibly because of sporadic reports that PS siRNAs
Maria Peiris-Pagès et al.
Aging, 11(14), 4801-4835 (2019-07-18)
Using proteomics analysis, we previously compared MCF7 breast cancer cells grown as 3D tumor spheres, with the same cell line grown as monolayers. Our results indicated that during 3D anchorage-independent growth, the cellular machinery associated with i) mitochondrial biogenesis and
Impaired oxidative phosphorylation regulates necroptosis in human lung epithelial cells
Koo MJ, et al.
Biochemical and Biophysical Research Communications, 464(3), 875-880 (2015)

Related Content

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Questions

1–10 of 10 Questions  
  1. Can I use R0278 for human protein extract for lymphocytes?

    1 answer
    1. This product has not been tested for protein extract from human lymphocytes. References using RIPA buffer with human lymphocytes can be found here:
      https://www.sciencedirect.com/science/article/pii/S0021925818639675
      https://www.sciencedirect.com/science/article/pii/S000887491530037X?via%3Dihub

      Helpful?

  2. Can I use the Ripa Buffer to extract yeast protein and make the Bradford assay on it?

    1 answer
    1. RIPA (Radio-Immunoprecipitation Assay) Buffer enables rapid, efficient cell lysis and solubilization of proteins from both adherent and suspension cultured mammalian cells. For more information, please see the Technical Bulletin: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/350/775/r0278bul.pdf

      Helpful?

  3. How long can the RIPA buffer be stored in 4C? I stored it for almost 6 years, can I still use it?

    1 answer
    1. Regarding stability in the recommended temperature of 2-8 °C, this product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  4. Can this be used for Western blot experiments?

    1 answer
    1. Yes, this product would be suitable for western blotting.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. How does Product R0278, RIPA buffer, compare to Product No. C2978 (CelLytic M)?

    1 answer
    1. RIPA buffer contains SDS, which is considered to be a harsh detergent; it can denature many proteins. C2978 is a buffered solution (pH 7.6) that contains a mild detergent; it is non-denaturing for most proteins.  The exact formulation of C2978, however, is proprietary.

      Helpful?

  7. Can Product R0278, RIPA buffer, be used on tissue samples, or only on cultured cells?

    1 answer
    1. R0278 can be used on tissue, if it is well homogenized.

      Helpful?

  8. I left the bottle of Product R0278, RIPA buffer, on my bench for about a week. Can I still use it?

    1 answer
    1. Yes, you should be able to use this product.  Although we recommend storage in the refrigerator at 2-8°C for long-term storage, Product No. R0278 may be kept at ambient temperature (our normal shipping condition) for a short time (approximately 2 weeks).

      Helpful?

  9. After preparing a lysate using Product R0278, RIPA Buffer, how should it be stored? And for how long can the lysate be stored?

    1 answer
    1. Lysates containing the appropriate inhibitor(s) can be stored at -70°C for up to a year.

      Helpful?

  10. Does Product R0278, RIPA buffer, contain any protease inhibitors or phosphatase inhibitors?

    1 answer
    1. This buffer does not contain any protease or phosphatase inhibitors, however, they may be added to insure against possible proteolysis.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service