Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX0737

Supelco

Toluene

OmniSolv®

Synonym(s):

Methylbenzene, FM approval

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-625-9

Agency

suitable for EPA 1613
suitable for EPA 1633

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

OmniSolv®

Assay

≥99.9% (toluene)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water (H2O)
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤3 ppm

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

solubility

0.52 g/L

density

0.865 g/mL at 25 °C (lit.)

UV cutoff

≤285 nm

UV absorption

λ: 285 nm Amax: ≤1.00
λ: 286 nm Amax: ≤0.70
λ: 288 nm Amax: ≤0.40
λ: 300 nm Amax: ≤0.10
λ: 350 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®
TX0737P-1 is packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (Toluene)99.9% minCapillary ECD responsive substances (as PCNB)5ng/L maxColor (APHA)10 maxEvaporation residue3ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardRefractive index (at 20°C)UV Abs. at 285 nm1.00AU maxUV Abs. at 286 nm0.70AU maxUV Abs. at 288 nm0.40AU maxUV Abs. at 300 nm0.10AU maxUV Abs. at 350 nm0.01AU maxUV Cut-off285nm maxWater (H2O)0.02% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service