Skip to Content
MilliporeSigma
Toluene JIS 1000, ≥99.8%

Toluene

Toluene JIS 1000, ≥99.8%
Synonym(s):
Toluene
Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique