Key Documents
TANKSDS2UV
Milli-Q
Loop UV Lamp Assembly 254 nm
Maintains consistent purity of stored water
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
packaging
pkg of 1 unit
parameter
254 nm (wavelength)
compatibility
for use with SDS
General description
This water purification accessory is designed for use with SDS 500 storage tanks. Pure water in the reservoir is distributed to multiple locations through the loop. This 254 nm UV lamp limits bacterial growth in the loop, thereby protecting the quality of pure water.
Application
This accessory prevents bacterial proliferation and biofilm buildup in the distribution loop.