Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ZFA100001

Milli-Q

A10® TOC Monitor

suitable for water monitoring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NA.21

packaging

pkg of 1 unit

application(s)

water monitoring

shipped in

ambient

General description

A10® TOC Monitor can be used for the accurate online measurement of total organic carbon (TOC) content in ultrapure water delivered by Milli-Q® water purification systems without a built-in TOC monitor. It is compatible with any high purity water laboratory system.

Application

A10® TOC Monitor is an ideal accessory for use in water monitoring. It is reported for use in the measurement of TOC/organic content in purified water and in water used as eluent in LC-MS studies.

Features and Benefits

  • High accuracy enabled by complete oxidation of organics to their end-point reaction, attributed to oxidation and conductivity measurements occurring within the same cell.
  • Range (1 – 999) and precision passing USP & EP suitability tests according to Milli-Q® Quality Assurance System standard procedures and specifications.
  • Traceability of measurement equipment is performed as per the Milli-Q® Calibration System. Certification of calibration is provided to comply with the industry requirements. A10® TOC Monitor can be recalibrated at user request and has been designed to take into account the suitability test requirements as specified in USP <643>.

Legal Information

A10 is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P W M H Smeets et al.
Water research, 40(17), 3239-3248 (2006-08-30)
To determine the disinfection efficacy of ozonation, water companies can apply several disinfection calculation methods. The goal of this study was to evaluate the use of the T10 and continuous stirred tank reactor (CSTR) method to extrapolate inactivation rates of
Cecilia Regnault et al.
Journal of chromatography. A, 1030(1-2), 289-295 (2004-03-27)
Improvements in trace enrichment techniques combined with the sensitivity of mass spectrometry offer enhanced opportunities to analyze ever lower concentrations of drugs, metabolites, pesticides or environmental pollutants. To perform HPLC and liquid chromatography-mass spectrometry (LC-MS) analyses under optimum conditions, the
Nikoloz Nioradze et al.
Analytical chemistry, 87(9), 4836-4843 (2015-04-07)
Highly oriented pyrolytic graphite (HOPG) is an important electrode material as a structural model of graphitic nanocarbons such as graphene and carbon nanotubes. Here, we apply scanning electrochemical microscopy (SECM) to demonstrate quantitatively that the electroactivity of the HOPG basal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service