Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

XX6200006P

Millipore

Filter forceps, blunt end, stainless steel

Synonym(s):

Forceps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104925
NACRES:
NB.24

material

302 stainless steel
polypropylene handle

Quality Level

Agency

suitable for EPA 1633
suitable for EPA OTM-45
suitable for ISO 21675 2019

application(s)

PFAS testing

shipped in

ambient

Features and Benefits

  • Beveled, unserrated tips to protect delicate membrane surfaces
  • It may be sterilized by autoclaving or flame sterilization
  • Use for gentle handling of membranes

Other Notes

Ensure your filtration membranes are not damaged while handling by using these blunt-ended forceps.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

For NIOSH 9000this page lists applicable products and materials to do the application

For NIOSH 9000this page lists applicable products and materials to do the application

The possible causes and potential remedies for challenges encountered as a result of blocking, washing, antibody incubation, and detection/exposure of Western blots

Protocols

This article is a protocol for Simple dialysis of small volumes using V Series membranes

The SNAP i.d. 2.0 Protein Detection System is the second generation of the SNAP i.d. method for detecting immunoreactive proteins on Western blots.

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service