Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SCC483

Sigma-Aldrich

HCC95 Human Lung Squamous Carcinoma Cell Line

Human

Synonym(s):

HCC95 cell line;Human lung carcinoma cell line;Lung squamous carcinoma cell line;Hamon Cancer Center 95;Human lung squamous carcinoma cell line;LUSC cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

HCC95 Human Lung Squamous Carcinoma Cell Line,

biological source

human

Quality Level

packaging

vial of ≥1X10⁶ cells

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable
cell migration: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

≥1x106 viable HCC95 Lung Squamous Carcinoma Cancer Cells: (Catalog No. SCC483). Store in liquid nitrogen.

Application

  • Each vial contains >_ 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for interspecies contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Lung cancer is the leading cause of cancer death, causing 23% of all cancer mortality in 2020. Among all lung cancer cases, 85% are categorized as non-small cell lung cancer (NSCLC) which is a heterogeneous group of diseases with poor prognosis and a 5-year survival rate of less than 20%. Among all NSCLC cases, 30-40% are categorized as lung squamous carcinoma (LUSC) which often affects the main airways and bronchi and is attributed to smoking as its principal etiology.

The HCC95 cell line is a human lung squamous cell carcinoma line, collected from a metastatic site of pleural effusion. Recent transcriptomic characterizations show that HCC95 show high similarity to the typical LUSC transcriptome.



Source
HCC95 human squamous lung carcinoma cells were derived from a 65-year-old male with NSCLC.

References
1.J Natl Compr Canc Netw. 2013, 11(6): 645–653 DOI: 10.6004/jnccn.2013.0084
2.Nat Rev Cancer 2014, 14(8): 535–546 DOI: 10.1038/nrc3775
3.J Thorac Dis. 2013, 5(Suppl 5): S463–S478 DOI: 10.3978/j.issn.2072-1439.2013.08.43
4.J Clin Oncol. 2010, 28(36): 5311–5320 DOI: 10.1200/JCO.2010.28.8126
5.Clin Can Res. 1999, 5(5): 991-1000
6.Nat Commun. 2019, 10(1): 3574 DOI: 10.1038/s41467-019-11415-2
7.J Cancer Res Clin Oncol 2010, 136(4): 493–502 DOI: 10.1007/s00432-009-0681-6

Features and Benefits

Among the NSCLC cell lines, HCC95 has been shown to retain the lung squamous carcinoma (LUSC) disease phenotype. Early research showed that HCC95 is very similar to the original tumor genetically and histochemically, with poor differentiation and formation of keratinization. Collectively, these demonstrate that HCC95 is a valuable cell line for modeling LUSC with high fidelity in vitro. A 2010 study has exemplified its use in studying NSCLC sensitivity to cisplatin treatment.

Storage and Stability

HCC95 cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service