Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SCC468

Sigma-Aldrich

T24T Human Bladder Carcinoma Cell Line

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

Product Name

T24T Human Bladder Carcinoma Cell Line,

biological source

human

Quality Level

packaging

vial of ≥1X10⁶ cells/vial mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell analysis: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

Bladder cancer originates from the epithelium that covers the inner surface of the bladder. The most common type is urothelial or transitional carcinoma (1). Most of the newly diagnosed cases (75%) are non-invasive, if the tumor becomes invasive it is more likely to metastasize to lymph nodes and other organs. Two common alterations have been identified in non-invasive tumors, deletion of chromosome 9 and a point mutation in FGFR3 (Fibroblast growth factor receptor 3) (2). Activating mutations in RAS gene family members are also common in bladder carcinoma and it is estimated that activation of the RAS-MAPK pathway may contribute to over 80% of the non-invasive cases (1). Loss of function of tumor suppressor genes like TP53 and RB1 are common in cases of invasive disease (1). After diagnosis the patient requires resection of the tumor and in cases of invasive disease chemotherapy is also provided.The original cell line T24 has been used to study bladder tumor biology but has some limitations because it lacks tumorigenicity when injected into immunocompromised mice. In contrast, T24T has increased metastatic ability when tested in vivo and expresses higher levels of HRAS mRNA (3). Increased HRAS activity leads to focal adhesion disassembly and los of CDH1 and b-catenin from intercellular junctions (3). Contact inhibition growth is reduced in T24T compared with T24 (3). Additionally, T24T does not inhibit its motile activity at higher density like T24 does.T24 cell line was generated from an invasive human transitional cell carcinoma of the bladder (4). T24T is a derivative of T24.1. Nat Rev Dis Primers 2017, 3: 17022.2. Int J Cancer 2006, 119(5): 1212-1215.3. Genes Chromosomes Cancer 2000, 27(3): 252-263.4. Int J Cancer 1973, 11(3): 765-773.
Each vial contains ≥1X10⁶ viable cells.Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services Cells are negative for mycoplasma contamination.

Features and Benefits

T24T human bladder cancer cell line is a useful model for the study of metastatic bladder cancer.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service