MilliporeSigma
HS9930
Cardboard Slide Tray
Holds 30 x slides, black, trim
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS9930
Feature:
Holds 30 x slides
Material:
cardboard
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
cardboard
feature
Holds 30 x slides
manufacturer/tradename
Heathrow Scientific HS9930
size
281x328x10 mm
color
black, trim
Features and Benefits
  • Easily organize slides with the index on the front cover that corresponds with the number grid inside
  • Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
  • Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
  • Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
  • Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
  • Disposable
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray
Choose from one of the most recent versions:
Certificates of Analysis (COA)