Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

MTRACEMC1

Millipore

M-TRACE® Touch Mobile Computer

M-TRACE® All-in-One PC, suitable for sterility testing

Synonym(s):

Mobile Device, Mobile computer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43210000
NACRES:
JA.12

packaging

pack of -1 unit

Quality Level

application(s)

pharmaceutical
sterility testing

compatibility

M-TRACE® All-in-One PC
M-TRACE® Mobile Computer Mount

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.


The M-TRACE® Touch Mobile Computer is a digital companion for sterility testing workflows. The mobile computer hosts the M-TRACE® Mobile App, which is part of the M-TRACE® Electronic Test Record Software. The device enables easy and efficient data capture on consumables and the addition of test results to the test record. The consumables′ data matrix codes can be scanned independently from any location. The data captured will automatically be pushed and added to the respective test records in the M-TRACE® All-in-One PC (MTRACEPC1). The M-TRACE® Touch Mobile Computer can be conveniently mounted outside the isolator/LFH pane with the M-TRACE® Mobile Computer Mount (MTRACEMM1). The mobile computer mount facilitates hands-free scanning and registration of the information on consumables while the operator′s hands are busy and occupied in the isolator. The M-TRACE® Touch Mobile Computer is clean room friendly and is compliant with the QC sterility testing environment requirements.

Application

M-TRACE® Touch Mobile Computer is used for easy and efficient data capture on consumables and the addition of test results to the test record during sterility testing and other quality control workflows.

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service