Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MTRACEHS1

Millipore

M-TRACE® Headset

for use with M-TRACE® All-in-One Computer, for use with Steritest® Symbio pump

Synonym(s):

Headset with mic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43210000
NACRES:
JA.12

packaging

pack of -1 unit

application(s)

pharmaceutical
sterility testing

compatibility

for use with M-TRACE® All-in-One Computer
for use with Steritest® Symbio pump

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.


M-TRACE® Headset is a cleanroom-friendly accessory of the M-TRACE® All-in-One Computer (MTRACEPC1 ). The M-TRACE™ Headset enables the voice control of the M-TRACE® Electronic Test Record Software and the Steritest® Symbio pump. Based on instructions from the software, the operator provides voice commands, and feedback is incorporated into the electronic test record. This real-time documentation can be achieved, despite the challenging work environment where the technician’s hands are often occupied in the microbiology QC testing lab e.g., inside an isolator.

Application

The M-TRACE® Headset enables the voice control of the M-TRACE® Electronic Test Record Software and the Steritest® Symbio pump to create or start test records and is used during sterility testing and other quality control workflows.

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service