Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAB5258

Sigma-Aldrich

Anti-Synaptophysin Antibody, clone SY38

clone SY38, Chemicon®, from mouse

Synonym(s):

synaptophysin, Synaptophysin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SY38, monoclonal

species reactivity

bovine, rat, avian, fish, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

General description

Synaptophysin is a 38 kDa glycoprotein present in the membrane of neuronal presynaptic vesicles in brain, spinal cord, retina, vesicles of adrenal medulla, neuromuscular junctions, and endocrine cells. It is also expressed by neuroendocrine cells throughout the body, both normal and neoplastic. Synaptophysin is a useful marker for the identification of normal neuroendocrine cells and neuroendocrine neoplasms. This antibody reacts with neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, and gastointestinal mucosa. It also reacts with a wide spectrum of neuroendocrine neoplasms of neural type including neuroblastomas, ganglioneuroblastomas, ganglioneuromas, pheochromocytomas, chromaffin, and non chromaffin paragangliomas.

Specificity

The antibody reacts with presynaptic vesicles of cerebral and spinal neurons, of neuromuscular endplates and with the retina of man, cow, rat and mouse. The antibody also reacts with vesicles of adrenal medulla and islet cells, and additionally allows specific staining of neuronal, adrenal and neuroepithelial tumors, such as pheochromocytoma, paraganglioma, islet cell tumors (incl. Insulinoma), medullary thyroid carcinoma and diverse pulmonary and gastro-intestinal carcinoids. The antibody also stains neurosecretory vesicles of certain culture cells, e.g. of the rat cell line PC-12.

Immunogen

Vesicular fraction of bovine brain. The SY38 epitope is to a pentapeptide repeat structure in the carboxy-terminal cytoplasmic tail of synaptophysin (Knaus & Betz 1990).

Application

Detect Synaptophysin using this Anti-Synaptophysin Antibody, clone SY38 validated for use in FC, IC, IH & WB.
Flow Cytometry (FACS):
A previous lot of this antibody was used in Flow Cytometry.

Western Blot:
(Provoda, 2000) Reacts with a 38 kDa transmembrane glycoprotein

Immunohistochemistry:
For relatively cytoplasm-rich neuroendocrinic tumors a final concentration of 1 µg/mL is recommended. Concerning cytoplasm deficient tumors a concentration of 2 µg/mL should be used. Ideal frozen sections (4-5 mm) are obtained from shock-frozen tissue samples. The frozen sections are air-dried and then fixed with acetone for 5-10 min at -20°C. Excess acetone is allowed to evaporate at room temperature. Material fixed in alcohol or formalin and embedded in paraffin can also be used.
It is advantageous to block unspecific binding sites by overlaying the sections with fetal calf serum for 20-30 min at room temperature. Excess of fetal calf serum is removed by decanting before application of the anti-body solution.
Cytocentrifuge preparations of single cells or cell smears are also fixed in acetone. These preparations should but should be followed directly by antibody treatment.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Routinely evaluated by Western Blot on mouse brain lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected synaptophysin on 10 μg of mouse brain lysates.

Target description

38 kDa

Linkage

Replaces: 04-1019

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1κ in buffer containing 20 mM sodium phosphate buffer, 0.25 M NaCl, containing 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8ºC from date of receipt. DO NOT FREEZE.

Analysis Note

Control
Positive Control: Pancreas tissue
Negative Control: Normal mouse serum
Rat hippocampus tissue, mouse brain lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Fuchs et al.
Neurobiology of disease, 70, 53-68 (2014-06-22)
Mutations in the X-linked cyclin-dependent kinase-like 5 (CDKL5) gene have been identified in a neurodevelopmental disorder characterized by early-onset intractable seizures, severe developmental delay, intellectual disability, and Rett's syndrome-like features. Since the physiological functions of CDKL5 still need to be
Hengchang Li et al.
Frontiers in neuroscience, 13, 706-706 (2019-07-30)
Children, after multiple exposures to general anesthesia, appear to be at an increased risk of developing learning disabilities. Almost all general anesthetics-including sevoflurane, which is commonly used for children-are potentially neurotoxic to the developing brain. Anesthesia exposure during development might
Sven Akkerman et al.
PloS one, 9(11), e111692-e111692 (2014-11-06)
Drug effects are usually evaluated in animals housed under maximally standardized conditions. However, it is assumed that an enriched environment (EE) more closely resembles human conditions as compared to maximally standardized laboratory conditions. In the present study, we examined the
Arielle R Strzelewicz et al.
Hormones and behavior, 111, 46-59 (2019-02-02)
Early life exposure to a low security setting, characterized by a scarcity of resources and limited food access, increases the risk for psychiatric illness and metabolic dysfunction. We utilized a translational rat model to mimic a low security environment and
Iwona Ksiazek et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(27), 7183-7195 (2007-07-06)
Agrin-deficient mice die at birth because of aberrant development of the neuromuscular junctions. Here, we examined the role of agrin at brain synapses. We show that agrin is associated with excitatory but not inhibitory synapses in the cerebral cortex. Most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service