Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MPSB-250ML

Millipore

mPAGE® 4X LDS Sample Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.56

form

liquid

packaging

vial of 250 mL

Looking for similar products? Visit Product Comparison Guide

General description

Used to prepare protein samples for denaturing polyacrylamide gel electrophoresis (PAGE) with mPAGE and most other Bis-Tris gels. LDS sample buffer contains lithium dodecyl sulfate with pH at 8.4 for optimal activity of the reducing agent.

  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

Western Blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service