Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST688

Basalt rock

NIST® SRM® 688

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in evaluating the accuracy of analytical methods and instruments used in the analysis of geological type materials. SRM 688 is a finely powdered basalt rock that was obtained from a Cenozoic basalt flow near Jaclpot, Nevada. For more information, please refer to the SDS and COA.

SRM 688_cert

SRM 688_SDS

Application

The SRM is intended for the assessment of the accuracy of instruments and analytical methods used in the analysis of geological type materials.

Other Notes

Certified for the analytes listed below.
Aluminum oxide (Al2O3), Iron(II) oxide (FeO), Iron(III) oxide (Fe2O3), Potassium oxide (K2O), Manganese(II) oxide (MnO), Sodium oxide (Na2O), Phosphorus Pentoxide (P2O5), Silicon dioxide (SiO2), Titanium dioxide (TiO2)

Reference/Informational value is provided for the following analytes.
Calcium oxide (Cao), Carbon dioxide (CO2), Fluorine (F), Magnesium oxide (MgO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service