Skip to Content
MilliporeSigma
All Photos(1)

Documents

MMHE-44K

Millipore

MILLIPLEX® Mouse Metabolic Hormone Expanded Panel - Metabolism Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Metabolic syndrome is a cluster of conditions that occur together; it includes increased blood pressure, elevated insulin levels, excess body fat around the waist and abnormal cholesterol levels.Key features of metabolic syndrome include insulin resistance, glucose intolerance, hypertension, dyslipidemia and central obsesity- all of which are risk factors for atherosclerosis, coronary heart disease, type 2 diabetes, kidney disease, and even premature death.Adults with metabolic syndrome show a low-grade inflammation, whose link with obesity may be dysregulated adipocyte production of pro- and anti-inflammatory factors. Consequently, research done in this area covers multi-faceted fields of cytokines, acute phase proteins, diabetes, and obesity related hormones, as well as other cardiovascular disease biomarkers.The MILLIPLEX® Mouse Metabolic Hormone Expansion Panel is a 15-plex kit to be used for the simultaneous quantification of any or all of the following analytes in tissue/lysate and culture supernatant samples and mouse serum or plasma: Amylin Active, C-Peptide 2, Ghrelin, GIP, GLP-1 Active or GLP-1 Total, Glucagon, IL-6, Insulin, Leptin, MCP-1, PP, PYY, Resistin, Secretin, and TNFα.Note: GLP-1 Active an GLP-1 Total cannot be run together in the same assay.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The Mouse Metabolic Hormone Expanded Panel, using the Luminex® xMAP technology, enables the simultaneous analysis of 15 protein biomarkers in mouse serum, plasma and tissue/lysate and culture supernatant samples.There are 16 biomarkers to choose from in this kit. However, this is a 15-plex kit since the following analytes cannot be run simultaneously: GLP-1 (Active) & GLP-1 (Total).The following analytes require the addition of protease inhibitors to the serum or plasma samples (see sample collection and storage section in the protocol for details): GLP-1 (DPPIV inhibitor); Amylin (protease inhibitor cocktail); Glucagon (Aprotinin); Ghrelin (Active) (serine protease inhibitor). The protocol recommends serum or plasma samples be run neat. Tissue culture samples may be run neat or may require a dilution with an appropriate control medium. For optimal sensitivity, use an overnight (16 - 18 hour) incubation. Each well requires 10 uL of sample, standard or control.Analytes available:Amylin (Active);C-Peptide 2;Ghrelin (Active);GIP (Total);GLP-1 (Active);GLP-1 (Total);Glucagon;IL-6;Insulin;Leptin;MCP-1/CCL2;Pancreatic Polypeptide (PP);PYY (total);Resistin;Secretin;TNF-α

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

New xMAP® Sheath Fluid PLUS eliminates microbial resistance, enhancing MILLIPLEX® multiplex assays.

New xMAP® Sheath Fluid PLUS eliminates microbial resistance, enhancing MILLIPLEX® multiplex assays.

New xMAP® Sheath Fluid PLUS eliminates microbial resistance, enhancing MILLIPLEX® multiplex assays.

New xMAP® Sheath Fluid PLUS eliminates microbial resistance, enhancing MILLIPLEX® multiplex assays.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service