Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MERS03SAP

Millipore

Millicell® ERS 3.0 Standard Electrode

Standard adjustable Electrode for Millicell ERS 3.0 Digital Voltohmmeter. Can be used with 6-, 12-, 24-well inserts.

Synonym(s):

ERS, Standard electrode, TEER Reader electrode, TEER Reader probe, Transepithelial Electrical resistance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.24

product line

Millicell®

General description

The Millicell® ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The Standard Electrode Probe fits into 3 different sizes of membrane inserts and receiver plates (6-well, 12-well, and 24-well). The Standard Electrode Probe is specially designed to be adjustable so that the depth of the electrode reaches the bottom of the well plate while the adjustable ring rests on top of the well. An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage. The small size of the electrodes enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes.

Application

To study endothelial/epithelial barrier models (like the blood-brain barrier models, gastrointestinal tract models, and pulmonary models), To study efficacy of drugs, To evaluate pharmacological toxicities

Features and Benefits

  • Ergonomic standing in-well probe
  • Adjustable electrode, compatible with a wide variety of cell culture inserts and plates
  • Built-in sensor to measure and record media temperature
  • Resistance range of 0 – 100 kΩ, with 1 Ω resolution
  • Demonstrates low noise, greater resolution, and high accuracy

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Related Content

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

Culture in 2.5D using small intestine duodenum organoids and Millicell® hanging cell culture inserts to monitor long-term drug side effects and toxicity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service