Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MERS03BAT

Millipore

Battery for Millicell® ERS 3.0

External battery for Millicell ERS 3.0 Digital Voltohmmeter

Synonym(s):

TEER Reader battery, TEER Reader external battery

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

product line

Millicell®

General description

The Millicell® ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The Millicell®ERS 3.0 system battery is a rechargable lithium-ion battery 14.4V/3.45Ah/49.70Wh. Each instrument is battery compatible but can run off only AC wall power in the absence of a battery.

Application

To study endothelial/epithelial barrier models (like the blood-brain barrier models, gastrointestinal tract models, and pulmonary models), To study efficacy of drugs, To evaluate pharmacological toxicities

Features and Benefits

  • Fast charging Lithium-ion battery can hold charge for several hours
  • Allows on the bench or in the hood use of Millicell ERS 3.0 system, without corded power

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Related Content

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service