Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MERS0396P

Millipore

Millicell® ERS 3.0 96-well Electrode

Specialized 96-well probe for Millicell® ERS 3.0 Voltohmmeter.

Synonym(s):

96-well electrode, TEER Reader 96-well probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.24

product line

Millicell®

General description

The Millicell® ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The 96-well electrode probe is designed for 96-well membrance inserts and receiver plates. The 96-well Electrode Probe is specially designed to be adjustable so that the depth of the electrode reaches the bottom of the well plate while the adjustable ring rests on top of the well. An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage. The small size of the electrodes enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes.

Application

To study endothelial/epithelial barrier models (like the blood-brain barrier models, gastrointestinal tract models, and pulmonary models), To study efficacy of drugs, To evaluate pharmacological toxicities

Features and Benefits

  • Ergonomic standing in-well probe
  • Adjustable electrode, compatible with a wide variety of cell culture inserts and plates
  • Built-in sensor to measure and record media temperature
  • Demonstrates low noise, greater resolution, and high accuracy

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Related Content

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service