Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MCSK10025

Millipore

Coli-Count Test Kit

The Coli-Count Blue Test Kit contains Blue Test Sampler (MC0010025) and Swab in Vial with Phosphate Buffer (MMSB10025)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.85

Quality Level

product line

Coli-Count

manufacturer/tradename

Coli-Count

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

suitability

coliforms

General description

The Swab Test Kit is a self-contained system, incorporating Coli-Count Samplers and units containing 18mL of sterile, phosphate buffer into which are fitted swab assemblies. The system is designed for use as swab tests for measuring coliform levels on flat or irregular surfaces. They can be used in a variety of applications in food production, food handling establishments, and other facilities to assess the efficiency of sanitation measures used to eradicate microbial contamination. The buffer units contain neutralizing agents to counteract the adverse effect of any residual chlorine or quaternary ammonium compounds that may be present on surfaces after sanitization. The kit contains 25 x dip tester + 25 x swabs

Application

The Coli-Count Swab Test Kit enables a fast and easy Coliform monitoring in food and beverages, dental unit water, dialysate, pharmaceuticals and cosmetics. This kit finds its application in Clinical Lab, Environmental Monitoring, Food and Beverage and Microbial monitoring

Analysis Note

Coliform colonies are blue or blue/green. Non-coliform colonies may be green, gray, or cream

Legal Information

Coli-Count is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service