Skip to Content
MilliporeSigma
All Photos(1)

Documents

MMSB10025

Millipore

Swab in vial with phosphate buffer

Neutralizing agents to counteract adverse effects of residual chlorine or quaternary ammonium compounds

Synonym(s):

Neutralizing buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
eCl@ss:
32160607
NACRES:
NB.24

material

Dacron swab(s)
polyethylene cap
polyethylene paddle
styrene-acrylonitrile (SAN) chamber

Quality Level

packaging

box of 25 units

input

sample type beverage(s)
sample type food(s)

application(s)

food and beverages
personnel monitoring
surface monitoring

compatibility

for use with Dip and Swab Samplers (MC0010025; MHPC10025; MY0010025)

shipped in

ambient

General description

The swab in the vial with phosphate buffer contains a neutralizing agent to counteract adverse effects of residual chlorine or quaternary ammonium compounds that may be present on surfaces after sanitization. The swab in the vial is used to sample the microbial levels on flat or irregular surfaces.

Application

Swab in the vial with phosphate buffer can be used in a variety of applications in food production, food handling establishments, and other facilities to assess the efficiency of sanitation measures used to eradicate microbial contamination. These swabs find their application in Clinical Lab, Environmental Monitoring, Food and Beverage, and Water Monitoring

Suitability

Environmental monitoring swab in phosphate buffer

Physical form

Color Code: Clear

Legal Information

Dacron is a trademark of Invista North America S.a.r.l.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service