Skip to Content
MilliporeSigma

1.02136

Contact C

Envirocheck, Contact Total Coliforms/E.Coli coated with Plate Count Agar and Chromocult Coliform Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41116116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

plastic cap, plastic slide, plastic tube

product line

Envirocheck

form

solid (ready-to-use)

manufacturer/tradename

Envirocheck

application(s)

food and beverages
surface monitoring
water monitoring

storage temp.

2-8°C

suitability

Escherichia coli (total count), coliforms

General description

(total coliforms/E.coli) Envirocheck
Envirocheck Contact Total Coliforms/E.Coli, a semi-quantitative detection of coliform bacteria on surfaces and in liquids, are flexible paddles that are coated with Plate Count Agar for simultaneous detection of coliforms and E. coli.Slide A is coated with Plate Count Agar to obtain the total viable (aerobic) count. Slide B is coated with Chromocult Coliform Agar for the simultaneous detection of coliforms and E. coli in water and food-samples.

Application

This medium is recommended for the simultaneous detection of coliforms and E. coli in water and food-samples. Envirocheck contact slides can be used for both surface and liquid testing in industrial and environmental monitoring.

Analysis Note

Count the number of colonies on both sides of the paddle. Plate Count Agar: Count the number of colonies for obtaining the total aerobic bacterial count (TVC). Chromocult Coliform Agar: Count the number of dark-blue to violet colonies (E. coli) and the number of salmon to red colonies for other coliforms. Additionally, E. coli can be confirmed by adding KOVAC′s Indole Reagent directly onto the colony (positive indole reaction: red colour).
Sterility: passes test
pH-value/side one (25 °C): 6.7 - 7.3
pH-value/side two (25 °C): 6.5 - 7.1
Growth/side one
- Escherichia coli ATCC 11775 (WDCM 00090):
good
- Escherichia coli O157:H7 ATCC 35150: good
- Citrobacter freundii ATCC 8090: good
- Salmonella enteritidis ATCC 13076 (WDCM 00030): good
Growth/side two
- Escherichia coli ATCC 11775 (WDCM 00090):
good
- Escherichia coli O157:H7 ATCC 35150: fair/good
- Citrobacter freundii ATCC 8090: good
- Salmonella enteritidis ATCC 13076 (WDCM 00030): good
Colonies
- Escherichia coli ATCC 11775 (WDCM 00090):
dark blue to violet
- Escherichia coli O157:H7 ATCC 35150: salmon to red
- Citrobacter freundii ATCC 8090: salmon to red
- Salmonella enteritidis ATCC 13076 (WDCM 00030): colourless
Incubation: 48 hrs.; 35 °C; aerobic.

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library