Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABS1935

Sigma-Aldrich

Anti-α-galactosidase A Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6G8, monoclonal

mol wt

calculated mol wt 48.77 kDa

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GLA(2717)

General description

a-Galactosidase A (UniProt: P06280; EC:3.2.1.22, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, Galactosylgalactosylglucosylceramidas GLA, Melibiase, Agalsidase) is encoded by the GLA gene (Gene ID: 2717) in human. a-galactosidase A is a homodimeric, lysosomal enzyme that catalyzes the hydrolysis of terminal alpha-galactosyl moieties from glycolipids and glycoproteins and participates to their degradation in the lysosome. It is synthesized with a signal peptide (aa 1-31), which is subsequently cleaved off to generate the mature form that is heavily glycosylated with asparagine-linked high mannose groups. Mutations in GLA gene have been linked to Fabry disease that is characterized by glycolipid accumulation in various tissues and leads to other complication such as renal failure and cardiac and cerebral complications.

Specificity

Clone 6G8 is a mouse monoclonal antibody that specifically detects human a-Galactosidase A.

Immunogen

Recombinant full-length human a-Galactosidase A (without the signal peptide).

Application

Quality Control Testing

Evaluated by Immunocytochemistry in Acetone fixed HEK 293 cells..

Immunocytochemistry Analysis: A 1:25 dilution of this antibody detected a-Galactosidase A in acetone fixed HEK 293 cells.

Tested Applications


Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected a-Galactosidase A in HeLa cells.

Immunohistochemistry Applications: A representative lot detected a-Galactosidase A in Immunohistochemistry applications (Tian, W., et. al. (2019). Nat Commun. 10(1):1785).

Immunocytochemistry Analysis: A representative lot detected a-Galactosidase A in acetone fixed HEK293 cells either HEK293 overexpressing GLA (A), HEK293 wild type (WT; B), or HEK293 cells with KO of GLA (C) (Courtesy of Asst. Prof. Zhang Yang, Ph.D., and Assoc. Prof. Ulla Mandel, Ph.D., Copenhagen Center for Glycomics, University of Copenhagen, Denmark).

Western Blotting Analysis: A representative lot detected a-Galactosidase A in lysates from HEK293 cells overexpressing GLA, HEK293 (WT), but not in HEK293 cells with GLA knockout (Courtesy of Asst. Prof. Zhang Yang, Ph.D., and Assoc. Prof. Ulla Mandel, Ph.D., Copenhagen Center for Glycomics, University of Copenhagen, Denmark).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-a-galactosidase A, clone 6G8, Cat. No. MABS1935, is a mouse monoclonal antibody that detects -a-galactosidase A and is tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service